Order Entry
Switzerland
ContactUsLinkComponent
 
VWR®, Taq DNA polymerase
 
undefined
VWR®, Taq DNA polymerase
 

 

  • Application:
    Standard amplification
  • Format:
    Enzyme with/without buffer
  • High fidelity:
    No
  • Hot start:
    No
  • Storage conditions:
    – 20 °C (long term), + 4 °C (6 months)
  • Amplicon size:
    ≤ 5kb
  • Processivity:
    Standard
  • Exonuclease activity:
    5’→3’
  • Product overhang:
    3'A
  • Features:
    Mg-free
  • Buffer composition:
    10x Key Buffer Mg-free and 25 mM MgCl2
  • Concentration:
    5 U/µl
  • Description:
    Taq DNA Polymerase (733-1313)
  • Size:
    5 x 0.1 ml + 10 x 1.5 ml
  • Units:
    2500

 

Frequently Bought Together