Level indicators, b.safe
About this item
Insert with screw-in thread for barrels or screw cap for canisters. With integrated level indicator to monitor the liquid level during filling as well as a neck with GL 45 thread for the assembly of a b.safe funnel with GL 45 thread.
- Prevent the waste canisters from overflowing
- Available for canisters and barrels
- Suitable for flammable liquid
The red visual pin goes up with increasing filling level. As soon as the visual pin fills the inspection glass completely, the canister is full and has to be exchanged.
Use of a b.safe funnel for filling of liquids and monitoring of the filling level at the same time to avoid run over.
8 Options Available Below
Product Details & Documents
Insert with screw-in thread for barrels or screw cap for canisters. With integrated level indicator to monitor the liquid level during filling as well as a neck with GL 45 thread for the assembly of a b.safe funnel with GL 45 thread.
- Prevent the waste canisters from overflowing
- Available for canisters and barrels
- Suitable for flammable liquid
The red visual pin goes up with increasing filling level. As soon as the visual pin fills the inspection glass completely, the canister is full and has to be exchanged.
Use of a b.safe funnel for filling of liquids and monitoring of the filling level at the same time to avoid run over.
Product Family Options
Product Information
- MaterialLengthForØAvailabilityPrice
- 590-0277Level indicator with GL 45 funnel connection, static dissipativeLevel indicator with GL 45 funnel connection, static dissipativePE—Female G2" / Tri-Sure 2"— mmLog in for availability1 * 1 STSFr. 384.00
- 590-0283Level indicator for barrelsLevel indicator for barrelsPTFE/PFA86 mmFemale G³/₄"23 mmLog in for availability1 * 1 STSFr. 709.00
- 590-0278Level indicator with GL 45 funnel connection, static dissipativeLevel indicator with GL 45 funnel connection, static dissipativePE—Female Mauser 2"— mmLog in for availability1 * 1 STSFr. 506.00
- 590-0282Level indicator for barrels
- 590-0284Level indicator with GL 45 funnel connection, static dissipativeLevel indicator with GL 45 funnel connection, static dissipativePE—Male S 60— mmLog in for availability1 * 1 STSFr. 494.00
- 590-0280Level indicator for barrelsLevel indicator for barrelsPE238 mmOpening Ø 50 - 100 mm50 mmLog in for availability1 * 1 STSFr. 968.00
- 590-0279Level indicator for barrelsLevel indicator for barrelsPE250 mmDairy pipe thread31 mmLog in for availability1 * 1 STSFr. 740.00
- 590-0281Level indicator for barrels, static dissipativeLevel indicator for barrels, static dissipativePE86 mmFemale G³/₄"23 mmLog in for availability1 * 1 STSFr. 289.00
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.