Orders LinkContactUsLinkComponent
Canisters, S90, b.safe
Canisters, S90, b.safe
Supplier:   Bohlender
Canisters, S90, b.safe
Supplier:  Bohlender
Certificates
Canisters, S90, b.safe
Canisters, S90, b.safe
2 Options Available Below
Product Details & Documents
Canister with thread S90 (DIN90) for the collection of liquid waste. Canister S90 in PE-EX with transparent stripe for observing the fill level.
  • Including screw cap
  • With UN-approval
  • b.safe accessories such as collecting trays, spouts and grounding cables, for dissipative canisters available
b.safe offers also suitable containers like flasks and canisters in addition to the wide range of caps for the extraction of solvents and waste caps or funnels for the collection of liquid waste.
Unless otherwise indicated, all b.safe canisters are supplied with UN-approval and can be used for the transport of hazardous goods.
Product Family Options
Product Information
  • Capacity
    Thread
    W×D×H
    Availability
    Price
  • 590-0303
    Canister
    Canister
    10,0 L
    S90
    195×195×370 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 35.60
    SFr. 35.60
    1 * 1 ST
       SFr. 35.60
  • 590-0304
    Canister, static dissipative
    Canister, static dissipative
    10,0 L
    S90
    195×195×370 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 143.00
    SFr. 143.00
    1 * 1 ST
       SFr. 143.00
Grounding cable
Product Information
  • Catalog #
    Description
    Length
    Availability
    Price
  • Grounding cable with clip and stainless steel strap
    1,5 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 91.40
    SFr. 91.40
    1 * 1 ST
       SFr. 91.40