Orders LinkContactUsLinkComponent
Thread adaptors, TYPE A, b.safe
Thread adaptors, TYPE A, b.safe
Supplier:   Bohlender
Thread adaptors, TYPE A, b.safe
Supplier:  Bohlender
Certificates
Thread adaptors, TYPE A, b.safe
Thread adaptors, TYPE A, b.safe
31 Options Available Below
Product Details & Documents
All adaptors of this type are supplied with a male thread, a female thread and recessed grips.
  • Stay flexible and independent
  • Recessed grips for an easy assembly
Your existing caps and waste caps do not fit with the flasks and canisters which you would like to use or vice versa? Simply use b.safe thread adaptors for a flexible combination of caps and waste caps independently from the thread on the flask and/or container.
Product Family Options
Product Information
  • Material
    Thread
    Availability
    Price
  • PE
    S 60/61 (male), S 50 (female)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 108.00
    SFr. 108.00
    1 * 1 ST
       SFr. 108.00
  • PE
    S 60/61 (male), S 51 (female)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 64.30
    SFr. 64.30
    1 * 1 ST
       SFr. 64.30
  • PE
    S 60/61 (male), S 71 (female)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 197.00
    SFr. 197.00
    1 * 1 ST
       SFr. 197.00
  • PP
    GL 45 (male), GL 32 (female)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 61.70
    SFr. 61.70
    1 * 1 ST
       SFr. 61.70
  • PP
    GL 45 (male), GPI 38-430 (female)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 60.00
    SFr. 60.00
    1 * 1 ST
       SFr. 60.00
  • PP
    GL 45 (male), 38×3 (female)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 74.20
    SFr. 74.20
    1 * 1 ST
       SFr. 74.20
  • PP
    GL 45 (male), S 40/GL 40 (female)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 59.40
    SFr. 59.40
    1 * 1 ST
       SFr. 59.40
  • PP
    GL 45 (male), S 51 (female)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 63.30
    SFr. 63.30
    1 * 1 ST
       SFr. 63.30
  • PP
    GL 45 (male), S 55 (female)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 62.80
    SFr. 62.80
    1 * 1 ST
       SFr. 62.80
  • PP
    GL 45 (male), S 60/61 (female)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 64.70
    SFr. 64.70
    1 * 1 ST
       SFr. 64.70
  • PP
    GL 45 (male), S 70/71 (female)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 119.00
    SFr. 119.00
    1 * 1 ST
       SFr. 119.00
  • PP
    GL 45 (male), G 1 ¹/₂" (female)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 62.90
    SFr. 62.90
    1 * 1 ST
       SFr. 62.90
  • PP
    S 51 (male), S 55 (female)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 66.50
    SFr. 66.50
    1 * 1 ST
       SFr. 66.50
  • PP
    S 51 (male), S 60/61 (female)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 67.00
    SFr. 67.00
    1 * 1 ST
       SFr. 67.00
  • PP
    S 55 (male), S 50 (female)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 63.10
    SFr. 63.10
    1 * 1 ST
       SFr. 63.10
  • PP
    S 55 (male), S 51 (female)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 65.80
    SFr. 65.80
    1 * 1 ST
       SFr. 65.80
  • PP
    S 55 (male), GL 45 (female)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 47.10
    SFr. 47.10
    1 * 1 ST
       SFr. 47.10
  • PP
    S 55 (male), S 60/61 (female)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 63.40
    SFr. 63.40
    1 * 1 ST
       SFr. 63.40
  • PP
    S 60/61 (male), S 51 (female)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 63.50
    SFr. 63.50
    1 * 1 ST
       SFr. 63.50
  • PP
    S 60/61 (male), S 55 (female)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 67.00
    SFr. 67.00
    1 * 1 ST
       SFr. 67.00
  • PP
    S 60/61 (male), B 63 (female)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 234.00
    SFr. 234.00
    1 * 1 ST
       SFr. 234.00
  • PP
    S 60/61 (male), S 65 (female)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 143.00
    SFr. 143.00
    1 * 1 ST
       SFr. 143.00
  • PP
    Tri-Sure 2" (male), G 2"/BSP 2" (female)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 61.10
    SFr. 61.10
    1 * 1 ST
       SFr. 61.10
  • PTFE
    S 40 (male), GL 45 (female)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 84.20
    SFr. 84.20
    1 * 1 ST
       SFr. 84.20
  • PTFE
    GL 45 (male), GL 32 (female)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 110.00
    SFr. 110.00
    1 * 1 ST
       SFr. 110.00
  • PTFE
    GL 45 (male), GPI 38-430 (female)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 88.00
    SFr. 88.00
    1 * 1 ST
       SFr. 88.00
  • PTFE
    GL 45 (male), GPI 38-400 (female)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 81.90
    SFr. 81.90
    1 * 1 ST
       SFr. 81.90
  • PTFE
    GL 45 (male), S 40/GL 40 (female)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 71.30
    SFr. 71.30
    1 * 1 ST
       SFr. 71.30
  • PTFE
    S 55 (male), S 40/GL 40 (female)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 113.00
    SFr. 113.00
    1 * 1 ST
       SFr. 113.00
  • PTFE
    S 55 (male), S 51 (female)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 91.50
    SFr. 91.50
    1 * 1 ST
       SFr. 91.50
  • PTFE
    S 55 (male), GL 45 (female)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 91.50
    SFr. 91.50
    1 * 1 ST
       SFr. 91.50