5-[3-(Trifluoromethyl)phenyl]-2-pyrimidinylhydrosulphide
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:344282-81-1
:
Formula:C₁₁H₇F₃N₂S
MDL Number:MFCD00243479
Melting Pt:262…265 °C
:256.25 g/mol
Storage Temperature:Raumtemperatur
Frequently Bought Together