5-Fluoro-o-anisidine 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1978-39-8
:
Formula:C₇H₈FNO
MDL Number:MFCD00077537
:141.15 g/mol
Boiling Pt:90 °C/0.76mmHg
Melting Pt:3 °C
Density:1.1981 g/cm³ (25 °C)
Flash Pt:108.9 °C
Storage Temperature:Ambient
UN:2810
ADR:6.1,III
:GHS07
Frequently Bought Together