7-Fluorobenzo-2-oxa-13-diazole-4-sulphonic acid ammonium salt
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:84806-27-9
:MFCD28385360
Formula:C₆H₆FN₃O₄S
MDL Number:MFCD02682109
:235.189 g/mol
Melting Pt:285…295 °C
Storage Temperature:Freezer
Frequently Bought Together