Order Entry
Switzerland
ContactUsLinkComponent
2-Bromo-2,2-dfluoroacetyl chloride 97%
Flammable
Corrosive
Irritant
2-Bromo-2,2-dfluoroacetyl chloride 97%
Catalog # APOSPC6436-5G
Supplier:  Apollo Scientific
CAS Number:  
Flammable
Corrosive
Irritant
2-Bromo-2,2-dfluoroacetyl chloride 97%
Catalog # APOSPC6436-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
Boiling Pt:50 °C
Storage Temperature:Refrigerator
UN:2924
ADR:3+8,III
 :GHS02
 :GHS05
 :GHS07
CAS Number:3832-48-2
Formula:C₂BrClF₂O
MDL Number:MFCD00069081