
AEBSF hydrochloride (4-(2-aminoethyl)benzenesulphonylfluoride hydrochloride) 98+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:30827-99-7
:
Formula:C₈H₁₁ClFNO₂S
MDL Number:MFCD00132962
:239.69 g/mol
Melting Pt:182…184 °C (dec.)
Storage Temperature:Freezer
UN:3261
ADR:8,II
:GHS05
Frequently Bought Together