3-(Trifluoromethyl)-o-anisidine 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:634187-08-9
:
Formula:C₈H₈F₃NO
MDL Number:MFCD08532489
:191.15 g/mol
Storage Temperature:Refrigerator
UN:2810
ADR:6.1,III
:GHS06
:GHS07
:GHS08
Frequently Bought Together