Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
[4-(Difluoromethyl)phenyl]methanol
[4-(Difluoromethyl)phenyl]methanol
[4-(Difluoromethyl)phenyl]methanol
APOSPC48346-250MG
:
Apollo Scientific
:
[4-(Difluoromethyl)phenyl]methanol
APOSPC48346-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
158.15 g/mol
:
GHS07
:
Ambient
:
444915-77-9
:
C₈H₈F₂O
:
MFCD20488855