
2-Fluoro-5-methylphenylboronic acid 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:166328-16-1
:
Formula:C₇H₈BFO₂
MDL Number:MFCD03428512
:153.9466 g/mol
Melting Pt:77…82 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together