Order Entry
Switzerland
ContactUsLinkComponent
1-(2-Tetrahydropyranyl)-3-(trifluoromethyl)-1H-pyrazole-5-boronic acid
1-(2-Tetrahydropyranyl)-3-(trifluoromethyl)-1H-pyrazole-5-boronic acid
Catalog # APOSPC448031-250MG
Supplier:  Apollo Scientific
CAS Number:  
1-(2-Tetrahydropyranyl)-3-(trifluoromethyl)-1H-pyrazole-5-boronic acid
Catalog # APOSPC448031-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :264.01 g/mol
Storage Temperature:Ambient
CAS Number:1141878-45-6
Formula:C₉H₁₂BF₃N₂O₃
MDL Number:MFCD17214248