7-(Trifluoromethyl)-1H-indole-2-carboxylic acid 95+%
Catalog # APOSPC446119-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:229.16 g/mol
Storage Temperature:Refrigerator
CAS Number:172216-98-7
Formula:C₁₀H₆F₃NO₂
MDL Number:MFCD02664424