
(3-Fluorophenyl)acetic acid 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:331-25-9
:
Formula:(C₆H₄F)CH₂COOH
MDL Number:MFCD00004331
:154.14 g/mol
Boiling Pt:151 °C/0.8mmHg
Melting Pt:42…48 °C
Flash Pt:113 °C
Storage Temperature:Ambient
EINECS:206-360-7
:GHS07
Frequently Bought Together