Order Entry
Switzerland
ContactUsLinkComponent
2-Chloro-6-fluoro-5-methylphenylboronic acid 97%
2-Chloro-6-fluoro-5-methylphenylboronic acid 97%
Catalog # APOSPC412003-5G
Supplier:  Apollo Scientific
CAS Number:  
2-Chloro-6-fluoro-5-methylphenylboronic acid 97%
Catalog # APOSPC412003-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :188.3917 g/mol
Storage Temperature:Ambient
CAS Number:352535-86-5
Formula:C₇H₇BClFO₂
MDL Number:MFCD03701535