4-Fluoro-3-iodotoluene 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:452-82-4
:
Formula:C₇H₆FI
MDL Number:MFCD00079759
Boiling Pt:122…125 °C/30mmHg
Density:1.833 g/cm³ (25 °C)
Molecular Weight:236.03 g/mol
Flash Pt:60 °C
Storage Temperature:Ambient
UN:1992
ADR:3+6.1 III
:GHS02
:GHS06
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together