Order Entry
Switzerland
ContactUsLinkComponent
1-Cyclopropyl-6-fluoro-7-chloro-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylic acid
Irritant
1-Cyclopropyl-6-fluoro-7-chloro-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylic acid
Catalog # APOSPC301100-100G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1-Cyclopropyl-6-fluoro-7-chloro-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylic acid
Catalog # APOSPC301100-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :282.6600 g/mol
Melting Pt:222…226 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:100361-18-0
Formula:C₁₂H₈ClFN₂O₃
MDL Number:MFCD08458865