
5-Fluoro-2-methylphenylboronic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:163517-62-2
:
Formula:C₇H₈BFO₂
MDL Number:MFCD03095047
Melting Pt:148…149 °C
:153.95 g/mol
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together