Order Entry
Switzerland
ContactUsLinkComponent
Methyl-2-methyltetrahydrofuran-2-carboxylate 95%
Methyl-2-methyltetrahydrofuran-2-carboxylate 95%
Catalog # APOSOR965309-1G
Supplier:  Apollo Scientific
CAS Number:  
Methyl-2-methyltetrahydrofuran-2-carboxylate 95%
Catalog # APOSOR965309-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :144.17 g/mol
Storage Temperature:Ambient
CAS Number:1218915-91-3
Formula:C₇H₁₂O₃
MDL Number:MFCD18651757