


Benzoic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:65-85-0
:
Formula:C₇H₆O₂
MDL Number:MFCD00002398
:122.12 g/mol
Boiling Pt:249 °C
Melting Pt:121…125 °C
Density:1.32 g/cm³ (25 °C)
Flash Pt:121 °C
EINECS:200-618-2
:GHS05
:GHS07
:GHS08
Storage Temperature:Ambient
Frequently Bought Together