Methyl-4-bromo-3-methoxybenzoate 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:17100-63-9
:
Formula:C₉H₉BrO₃
MDL Number:MFCD08436018
:245.07 g/mol
Melting Pt:58…61 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together