Order Entry
Switzerland
ContactUsLinkComponent
Ac-Ile-NHMe 95%
Ac-Ile-NHMe 95%
Catalog # APOSOR947952-1G
Supplier:  Apollo Scientific
CAS Number:  
Ac-Ile-NHMe 95%
Catalog # APOSOR947952-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :186.255 g/mol
Storage Temperature:Ambient
CAS Number:32483-16-2
Formula:C₉H₁₈N₂O₂
MDL Number:MFCD00237495