(±)-1-Octen-3-ol 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:3391-86-4
:
Formula:C₈H₁₆O
MDL Number:MFCD00004589
:128.21 g/mol
Storage Temperature:Ambient
Frequently Bought Together
n-Hexane, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
Diethyl ether stabilised, EMPLURA®, Supelco®
Supplier: Merck
Pyridine, EMPLURA®, Supelco®
Supplier: Merck
Glycerine ≥99.5%, AnalaR NORMAPUR® ACS analytical reagent, redistilled
Supplier: VWR Chemicals
Water, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Diethyl ether ≥99.7% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Chlorobenzene ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
n-Heptane ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Acetone ≥99.7%, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
Pyridine, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier: Merck
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
Ethanol, GPR RECTAPUR®
Supplier: VWR Chemicals
n-Hexane, EMSURE® ACS for analysis, Supelco®
Supplier: Merck
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals