Order Entry
Switzerland
ContactUsLinkComponent
D-Valine ethyl ester hydrochloride 95%
D-Valine ethyl ester hydrochloride 95%
Catalog # APOSOR929780-5G
Supplier:  Apollo Scientific
CAS Number:  
D-Valine ethyl ester hydrochloride 95%
Catalog # APOSOR929780-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :181.66 g/mol
Storage Temperature:Ambient
CAS Number:73913-64-1
Formula:C₇H₁₅NO₂·HCl
MDL Number:MFCD00191043