Order Entry
Switzerland
ContactUsLinkComponent
2,3-Dimethyl-1H-indole-5-carboxylic acid 97%
Irritant
2,3-Dimethyl-1H-indole-5-carboxylic acid 97%
Catalog # APOSOR9226-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2,3-Dimethyl-1H-indole-5-carboxylic acid 97%
Catalog # APOSOR9226-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :189.22 g/mol
Melting Pt:239…242 °C
Storage Temperature:Refrigerator
UN:2811
ADR:6.1,III
 :GHS07
CAS Number:14844-73-6
Formula:C₁₁H₁₁NO₂
MDL Number:MFCD00464053