Order Entry
Switzerland
ContactUsLinkComponent
1-Methylazepan-4-ol 95%
1-Methylazepan-4-ol 95%
Catalog # APOSOR920530-1G
Supplier:  Apollo Scientific
CAS Number:  
1-Methylazepan-4-ol 95%
Catalog # APOSOR920530-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :129.2000 g/mol
Storage Temperature:Ambient
CAS Number:19065-49-7
Formula:C₇H₁₅NO
MDL Number:MFCD00190597