Order Entry
Switzerland
ContactUsLinkComponent
2-Methyl-3-heptanone 95%
2-Methyl-3-heptanone 95%
Catalog # APOSOR918444-5G
Supplier:  Apollo Scientific
CAS Number:  
2-Methyl-3-heptanone 95%
Catalog # APOSOR918444-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Butyl Isopropyl ketone, 2-Methylheptan-3-one
 :
 :128.21 g/mol
Storage Temperature:Ambient
CAS Number:13019-20-0
Formula:C₈H₁₆O
MDL Number:MFCD00009481