Order Entry
Switzerland
ContactUsLinkComponent
3-Phenoxypropan-1-ol 95%
3-Phenoxypropan-1-ol 95%
Catalog # APOSOR917493-100G
Supplier:  Apollo Scientific
CAS Number:  
3-Phenoxypropan-1-ol 95%
Catalog # APOSOR917493-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :152.193 g/mol
Storage Temperature:Ambient
CAS Number:6180-61-6
Formula:C₉H₁₂O₂
MDL Number:MFCD00039549