Order Entry
Switzerland
ContactUsLinkComponent
 
cis-2-Phenyl-1,3-dioxan-5-ol 95%
  APOSOR906811-25G
 :  
cis-2-Phenyl-1,3-dioxan-5-ol 95%
  APOSOR906811-25G
 :  

 

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

 

 

 :
 :180.2000 g/mol
 :Ambient
 :4141-19-9
 :C₁₀H₁₂O₃
 :MFCD00274343