Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-(Trimethylsilyl)-2-propyn-1-ol
3-(Trimethylsilyl)-2-propyn-1-ol 98%
3-(Trimethylsilyl)-2-propyn-1-ol 98%
APOSOR900149-100G
:
Apollo Scientific
:
3-(Trimethylsilyl)-2-propyn-1-ol 98%
APOSOR900149-100G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
100 g
Seq:
3401
Environmentally Preferable:
:
3-(Trimethylsilyl)propargyl alcohol
:
:
128.25 g/mol
:
Ambient
:
5272-36-6
:
C₆H₁₂OSi
:
MFCD00002913