2',6'-Dimethoxyacetophenone
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: 1-(2,6-Dimethoxyphenyl)ethan-1-one, 1-(2,6-Dimethoxyphenyl)ethanone
- CAS Number:2040-04-2
- Formula:C₁₀H₁₂O₃
- MDL Number:MFCD00008729
- Molecular Weight:180.2 g/mol
- Storage Temperature:Ambient
5 Options Available Below
Product Details & Documents
Synonyms: 1-(2,6-Dimethoxyphenyl)ethan-1-one, 1-(2,6-Dimethoxyphenyl)ethanone
- CAS Number:2040-04-2
- Formula:C₁₀H₁₂O₃
- MDL Number:MFCD00008729
- Molecular Weight:180.2 g/mol
- Storage Temperature:Ambient
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product



