
4-Hydrazinobenzoic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:619-67-0
:
Formula:C₇H₈N₂O₂
MDL Number:MFCD00007581
:152.15 g/mol
Melting Pt:217…219 °C (dec.)
EINECS:210-609-5
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together