
Tetrahydropyran-3-methanol
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:14774-36-8
:
Formula:C₆H₁₂O₂
MDL Number:MFCD08235011
:116.1583 g/mol
Boiling Pt:81 °C/4mmHg
Density:1.0553 g/cm³ (25 °C)
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together