1-[4-(Bromomethyl)phenyl]methanamine
Catalog # APOSOR6190-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:200.08 g/mol
Storage Temperature:Ambient
CAS Number:769057-30-9
Formula:C₈H₁₀BrN
MDL Number:MFCD03410966