
1-(4-Chlorophenyl)-1-methylethylamine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:17797-11-4
:
Formula:C₉H₁₂ClN
MDL Number:MFCD08669636
:169.66 g/mol
Boiling Pt:91 °C/4mmHg
Storage Temperature:Ambient
UN:2735
ADR:8,III
:GHS05
Frequently Bought Together