Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Caproic acid 99%

Caproic acid 99%

Supplier:   Apollo Scientific
Caproic acid 99%
Supplier:  Apollo Scientific
Certificates
Caproic acid 99%

2 Options Available Below

Product Details & Documents
Synonyms:  Hexanoic acid
  • CAS Number:142-62-1
  • Formula:C₆H₁₂O₂
  • MDL Number:MFCD00004421
  • Molecular Weight:116.16 g/mol
  • Boiling Pt:202…203 °C
  • Melting Pt:–3 °C
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - SFr. 34.70
    SFr. 34.70
    1 * 25 g
       SFr. 34.70

  • Bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - SFr. 83.20
    SFr. 83.20
    1 * 100 g
       SFr. 83.20

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.