
2-(2-Methoxyethoxy)acetyl chloride 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:16024-55-8
:
Formula:C₅H₉ClO₃
MDL Number:MFCD02094014
:152.5762 g/mol
Boiling Pt:71…75 °C/10mmHg
Storage Temperature:Ambient
UN:3265
ADR:8,II
:GHS05
Frequently Bought Together