
7-Methyl-1H-indole-2-carboxylic acid
Catalog # APOSOR5661-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:175.19 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:18474-60-7
Formula:C₁₀H₉NO₂
MDL Number:MFCD00047168