3-Phenyl-2-propyn-1-ol 98+%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 3-Phenylprop-2-yn-1-ol
CAS Number:1504-58-1
:
Formula:C₉H₈O
MDL Number:MFCD00040914
:132.16 g/mol
Boiling Pt:129…130 °C/10mmHg
Density:1.06 g/cm³ (25 °C)
Flash Pt:>110 °C
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together