


2-Bromophenol 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:95-56-7
:
Formula:C₆H₅BrO
MDL Number:MFCD00002146
:173.01 g/mol
Boiling Pt:194…195 °C
Melting Pt:44380 °C
Density:1.49 g/cm³ (25 °C)
Flash Pt:42 °C
Storage Temperature:Ambient
EINECS:202-432-7
UN:1993
ADR:3,III
:GHS02
:GHS07
:GHS09
Frequently Bought Together