2-Hydroxy-3-nitro-6-picoline
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 6-Methyl-3-nitro-2(1H)-pyridinone, 2-Hydroxy-6-methyl-3-nitropyridine, 6-Hydroxy-5-nitro-2-picoline, 6-Methyl-3-nitropyridin-2-ol, 6-Methyl-3-nitro-2-pyridone, 2-Methyl-5-nitro-6-pyridone, 6-Hydroxy-2-methyl-5-nitropyridine
CAS Number:39745-39-6
:
Formula:C₆H₆N₂O₃
MDL Number:MFCD03001414
:154.13 g/mol
Melting Pt:222…226 °C
:GHS05
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together