5-Iodo-o-anisidine 98%
Catalog # APOSOR51812-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:249.05 g/mol
Melting Pt:85…87 °C
:GHS07
Storage Temperature:Ambient
CAS Number:77770-09-3
Formula:C₇H₈INO
MDL Number:MFCD03095407