
2-Methyl-1,8-naphthyridine-3-carboxylic acid 95+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:387350-60-9
:
Formula:C₁₀H₈N₂O₂
MDL Number:MFCD00202882
:188.19 g/mol
Melting Pt:191…195 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together