1,5-Anhydro-D-sorbitol
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 1-Deoxy-D-glucose, 1,5-Anhydro-D-glucitol
CAS Number:154-58-5
:
Formula:C₆H₁₂O₅
MDL Number:MFCD00067386
:164.16 g/mol
Melting Pt:142…143 °C
EINECS:205-829-3
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together