Order Entry
Switzerland
ContactUsLinkComponent
Methyl-4-chloro-7-methoxyquinoline-6-carboxylate 95%
Methyl-4-chloro-7-methoxyquinoline-6-carboxylate 95%
Catalog # APOSOR471642-25G
Supplier:  Apollo Scientific
CAS Number:  
Methyl-4-chloro-7-methoxyquinoline-6-carboxylate 95%
Catalog # APOSOR471642-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :251.6700 g/mol
Storage Temperature:Ambient
CAS Number:205448-66-4
Formula:C₁₂H₁₀ClNO₃
MDL Number:MFCD17016133