

2-Chloro-5-nitropyrimidin-4-amine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1920-66-7
:
Formula:C₄H₃ClN₄O₂
MDL Number:MFCD00127771
:174.544 g/mol
Melting Pt:221…226 °C
:GHS07
:GHS08
Storage Temperature:Ambient
Frequently Bought Together