6-Chloro-m-anisidine hydrochloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:85006-21-9
:
Formula:C₇H₉Cl₂NO
MDL Number:MFCD00012962
:194.06 g/mol
Melting Pt:207 °C (dec.)
EINECS:285-050-3
:GHS07
Storage Temperature:Ambient
Frequently Bought Together