Order Entry
Switzerland
ContactUsLinkComponent
2-(Ethoxycarbonyl)-1H-indole-3-carboxylic acid 97%
2-(Ethoxycarbonyl)-1H-indole-3-carboxylic acid 97%
Catalog # APOSOR43534-250MG
Supplier:  Apollo Scientific
CAS Number:  
2-(Ethoxycarbonyl)-1H-indole-3-carboxylic acid 97%
Catalog # APOSOR43534-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :233.23 g/mol
Melting Pt:262…263 °C
Storage Temperature:Ambient
CAS Number:441800-93-7
Formula:C₁₂H₁₁NO₄
MDL Number:MFCD18803524