2-Chloro-5-methylphenylboronic acid 96%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:193353-35-4
:
Formula:C₇H₈BClO₂
MDL Number:MFCD06659822
:170.40 g/mol
Melting Pt:203…205 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together